Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419377.6 | XP_419377.1 | 2 - 3 | 141 | 150 | |
GenBank mRNA | BX932118.1 | - | 2 - 3 | 90 | 150 | |
BX934462.2 | - | 2 - 3 | 89 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |