Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015290869.1 | XP_015146355.1 | 2 - 3 | 63 | 68 | |
GenBank mRNA | BX934727.2 | - | 2 - 3 | 59 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |