Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX929565.1 | - | 3 - 4 | 392 | 71 | |
CR385154.1 | - | 2 - 3 | 222 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |