Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277371.1 | NP_001264300.1 | 6 - 7 | 740 | 94 | |
GenBank mRNA | BU248955.1 | - | 4 - 5 | 493 | 94 | |
BU398630.1 | - | 1 - 2 | 84 | 94 | ||
CR387675.1 | - | 6 - 7 | 727 | 94 | ||
CR389655.1 | - | 2 - 3 | 262 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |