Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420181.6 | XP_420181.1 | 6 - 7 | 1038 | 63 | |
GenBank mRNA | CR407240.1 | - | 6 - 7 | 1032 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |