Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417733.6 | XP_417733.1 | 5 - 6 | 626 | 118 | |
GenBank mRNA | BX932284.1 | - | 3 - 4 | 431 | 118 | |
BX935491.2 | - | 5 - 6 | 587 | 118 |
Gene Symbol: RAB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143031.1 | XP_024998799.1 | 1 - 2 | 173 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |