Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935332.3 | XP_004935389.1 | 2 - 3 | 631 | 124 | |
XM_419430.5 | XP_419430.1 | 2 - 3 | 228 | 124 | ||
GenBank mRNA | BX935159.2 | - | 1 - 2 | 605 | 124 | |
CR353202.1 | - | 2 - 3 | 217 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |