Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281969.1 | XP_015137455.1 | 2 - 3 | 250 | 97 | |
GenBank mRNA | CR353752.1 | - | 2 - 3 | 234 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |