Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277841.1 | NP_001264770.1 | 7 - 8 | 752 | 76 | |
XM_015293127.2 | XP_015148613.1 | 7 - 8 | 924 | 76 | ||
GenBank mRNA | AJ396688.1 | - | 5 - 6 | 564 | 76 | |
BU334051.1 | - | 1 - 2 | 60 | 76 | ||
BX934554.2 | - | 7 - 8 | 752 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |