Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JKAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277554.1 | NP_001264483.1 | 6 - 7 | 764 | 79 | |
GenBank mRNA | BU317493.1 | - | 3 - 4 | 363 | 79 | |
BX936131.2 | - | 6 - 7 | 737 | 79 | ||
CV860445.1 | - | 4 - 5 | 646 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |