Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942966.3 | XP_004943023.1 | 20 - 21 | 2117 | 90 | |
XM_422119.6 | XP_422119.4 | 19 - 20 | 2092 | 90 | ||
GenBank mRNA | BX950354.2 | - | 10 - 11 | 1033 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |