Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282235.2 | NP_001269164.1 | 7 - 8 | 1449 | 70 | |
GenBank mRNA | CR385344.1 | - | 3 - 4 | 418 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |