Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282541.2 | XP_015138027.2 | 56 - 57 | 8982 | 137 | |
XM_015282543.2 | XP_015138029.2 | 55 - 56 | 8814 | 137 | ||
XM_025147818.1 | XP_025003586.1 | 56 - 57 | 8985 | 137 | ||
XM_025147819.1 | XP_025003587.1 | 55 - 56 | 8873 | 137 | ||
XM_025147820.1 | XP_025003588.1 | 56 - 57 | 8982 | 137 | ||
XM_025147821.1 | XP_025003589.1 | 52 - 53 | 8348 | 137 | ||
XM_025147822.1 | XP_025003590.1 | 55 - 56 | 8946 | 137 | ||
XM_025147824.1 | XP_025003592.1 | 55 - 56 | 8817 | 137 | ||
GenBank mRNA | CR385783.1 | - | 6 - 7 | 848 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |