Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277506.1 | NP_001264435.1 | 3 - 4 | 71 | ||
GenBank mRNA | BX933486.2 | - | 3 - 4 | 344 | 71 | |
BX935618.1 | - | 3 - 4 | 346 | 71 | ||
CR352574.1 | - | 3 - 4 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |