Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143900.1 | XP_024999668.1 | 362 | 82 | ||
XM_417970.6 | XP_417970.4 | 397 | 82 | |||
GenBank mRNA | CR407416.1 | - | 403 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus |