Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006425.1 | NP_001006425.1 | 4 - 5 | 454 | 90 | |
XM_015295400.2 | XP_015150886.1 | 4 - 5 | 585 | 90 | ||
GenBank mRNA | AJ719480.1 | - | 4 - 5 | 454 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |