Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204643.1 | NP_989974.1 | 3 - 4 | 387 | 103 | |
XM_015281284.2 | XP_015136770.1 | 4 - 5 | 458 | 103 | ||
XM_015281285.2 | XP_015136771.1 | 3 - 4 | 580 | 103 | ||
GenBank mRNA | AB005618.1 | - | 3 - 4 | 387 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |