Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NREP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205391.1 | NP_990722.1 | 2 - 3 | 139 | 86 | |
GenBank mRNA | U30520.1 | - | 2 - 3 | 139 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |