Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1MSW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205490.1 | NP_990821.1 | 1056 | 76 | ||
GenBank mRNA | M92038.1 | - | 1056 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Gallus gallus |