Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012794.1 | NP_001012812.1 | 6 - 7 | 1409 | 77 | |
XM_015275310.2 | XP_015130796.1 | 5 - 6 | 1316 | 77 | ||
GenBank mRNA | AJ720454.1 | - | 6 - 7 | 1409 | 77 | |
CR385883.1 | - | 4 - 5 | 814 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |