Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012861.1 | NP_001012879.1 | 2 - 3 | 229 | 79 | |
XM_015275856.2 | XP_015131342.1 | 2 - 3 | 715 | 79 | ||
XM_015275857.2 | XP_015131343.1 | 2 - 3 | 296 | 79 | ||
GenBank mRNA | AJ719502.1 | - | 2 - 3 | 229 | 79 | |
CR338760.1 | - | 2 - 3 | 204 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |