Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030637.1 | NP_001025808.1 | 6 - 7 | 696 | 150 | |
GenBank mRNA | AJ719602.1 | - | 6 - 7 | 696 | 150 | |
AJ851726.1 | - | 6 - 7 | 658 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |