Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001747.2 | NP_001001747.2 | 8 - 9 | 1542 | 77 | |
XM_004949029.3 | XP_004949086.1 | 9 - 10 | 1580 | 77 | ||
XM_004949031.3 | XP_004949088.1 | 7 - 8 | 4895 | 77 | ||
XM_004949032.3 | XP_004949089.1 | 7 - 8 | 1378 | 77 | ||
XM_015277412.2 | XP_015132898.1 | 8 - 9 | 4987 | 77 | ||
XM_015277413.2 | XP_015132899.1 | 7 - 8 | 1332 | 77 | ||
XM_015277414.2 | XP_015132900.1 | 7 - 8 | 1350 | 77 | ||
XM_015277415.2 | XP_015132901.1 | 6 - 7 | 4787 | 77 | ||
XM_015277416.1 | XP_015132902.1 | 6 - 7 | 568 | 77 | ||
GenBank mRNA | AJ704564.1 | - | 4 - 5 | 568 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |