Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205011.1 | NP_990342.1 | 2 - 3 | 176 | 95 | |
GenBank mRNA | AJ006405.1 | - | 2 - 3 | 176 | 95 | |
CR385723.1 | - | 2 - 3 | 177 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |