Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205314.1 | NP_990645.1 | 6 - 7 | 744 | 146 | |
XM_015288063.2 | XP_015143549.1 | 6 - 7 | 798 | 146 | ||
XM_015288064.2 | XP_015143550.1 | 6 - 7 | 834 | 146 | ||
GenBank mRNA | X16881.1 | - | 744 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |