Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030570.1 | NP_001025741.1 | 3 - 4 | 2248 | 103 | |
GenBank mRNA | AJ719785.1 | - | 3 - 4 | 2248 | 103 | |
CR386283.1 | - | 2 - 3 | 581 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |