Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030688.1 | NP_001025859.1 | 2 - 3 | 253 | 73 | |
XM_015279473.2 | XP_015134959.1 | 1 - 2 | 73 | |||
XM_015279474.2 | XP_015134960.1 | 2 - 3 | 73 | |||
GenBank mRNA | AJ851562.1 | - | 2 - 3 | 253 | 73 | |
CR338692.1 | - | 241 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |