Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030716.2 | NP_001025887.2 | 11 - 12 | 1204 | 80 | |
XM_015295755.2 | XP_015151241.1 | 11 - 12 | 1305 | 80 | ||
GenBank mRNA | AJ721005.1 | - | 11 - 12 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |