Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030949.2 | NP_001026120.2 | 2 - 3 | 254 | 70 | |
XM_015283013.2 | XP_015138499.1 | 3 - 4 | 415 | 70 | ||
XM_025147119.1 | XP_025002887.1 | 2 - 3 | 254 | 70 | ||
GenBank mRNA | AJ453193.1 | - | 2 - 3 | 255 | 70 | |
AJ721055.1 | - | 2 - 3 | 255 | 70 | ||
BU132419.1 | - | 2 - 3 | 185 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |