Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031288.1 | NP_001026459.1 | 7 - 8 | 1322 | 86 | |
GenBank mRNA | AJ720832.1 | - | 7 - 8 | 1322 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |