Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORCS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031357.1 | NP_001026528.1 | 3 - 4 | 432 | 79 | |
XM_015290581.2 | XP_015146067.1 | 3 - 4 | 489 | 79 | ||
XM_015290585.2 | XP_015146071.1 | 3 - 4 | 416 | 79 | ||
GenBank mRNA | AJ720525.1 | - | 3 - 4 | 432 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |