Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031408.1 | NP_001026579.1 | 2 - 3 | 109 | 74 | |
XM_015300078.2 | XP_015155564.1 | 2 - 3 | 272 | 74 | ||
GenBank mRNA | AJ720270.1 | - | 2 - 3 | 109 | 74 | |
CR389795.1 | - | 2 - 3 | 285 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |