Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005813.1 | NP_001005813.1 | 3 - 4 | 378 | 72 | |
GenBank mRNA | AJ719601.1 | - | 3 - 4 | 378 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |