Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006479.2 | NP_001006479.2 | 8 - 9 | 1215 | 124 | |
XM_015287342.2 | XP_015142828.1 | 9 - 10 | 1337 | 124 | ||
XM_015287343.2 | XP_015142829.1 | 9 - 10 | 1191 | 124 | ||
XM_025150725.1 | XP_025006493.1 | 11 - 12 | 6364 | 124 | ||
GenBank mRNA | AY313784.1 | - | 8 - 9 | 1215 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |