Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044689.1 | NP_001038154.1 | 5 - 6 | 924 | 89 | |
XM_025152263.1 | XP_025008031.1 | 5 - 6 | 746 | 89 | ||
GenBank mRNA | DQ010055.1 | - | 5 - 6 | 924 | 89 | |
EU165363.1 | - | 4 - 5 | 636 | 89 | ||
EU165364.1 | - | 4 - 5 | 636 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |