Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097537.1 | NP_001091006.1 | 1 - 2 | 139 | 144 | |
XM_015294250.2 | XP_015149736.1 | 2 - 3 | 431 | 144 | ||
XM_015294251.2 | XP_015149737.1 | 2 - 3 | 447 | 144 | ||
XM_015294252.2 | XP_015149738.1 | 2 - 3 | 395 | 144 | ||
GenBank mRNA | AM492777.1 | - | 1 - 2 | 139 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |