Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204841.2 | NP_990172.1 | 3 - 4 | 395 | 97 | |
GenBank mRNA | AF151968.2 | - | 3 - 4 | 395 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |