Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205520.4 | NP_990851.1 | 3 - 4 | 508 | 79 | |
GenBank mRNA | J02787.1 | - | 3 - 4 | 508 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |