Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204406.1 | NP_989737.1 | 1 - 2 | 431 | 144 | |
GenBank mRNA | AY225967.1 | - | 1 - 2 | 431 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Gallus gallus Probe spans exons |