Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147022.1 | XP_025002790.1 | 2 - 3 | 1316 | 97 | |
XM_025147023.1 | XP_025002791.1 | 2 - 3 | 917 | 97 | ||
XM_025147024.1 | XP_025002792.1 | 2 - 3 | 652 | 97 | ||
XM_025147025.1 | XP_025002793.1 | 2 - 3 | 901 | 97 | ||
XM_025147026.1 | XP_025002794.1 | 2 - 3 | 1753 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |