Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006233.1 | NP_001006233.1 | 10 - 11 | 1225 | 149 | |
GenBank mRNA | AJ720769.1 | - | 10 - 11 | 1225 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |