Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006261.1 | NP_001006261.1 | 4 - 5 | 591 | 138 | |
XM_025149742.1 | XP_025005510.1 | 4 - 5 | 525 | 138 | ||
GenBank mRNA | AJ720066.1 | - | 4 - 5 | 591 | 138 | |
AJ720312.1 | - | 4 - 5 | 572 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |