Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006503.1 | NP_001006503.1 | 4 - 5 | 401 | 119 | |
XM_015288626.2 | XP_015144112.1 | 2 - 3 | 578 | 119 | ||
XM_015288627.2 | XP_015144113.1 | 3 - 4 | 580 | 119 | ||
XM_015288629.2 | XP_015144115.1 | 2 - 3 | 585 | 119 | ||
XM_025151603.1 | XP_025007371.1 | 4 - 5 | 414 | 119 | ||
GenBank mRNA | AJ720183.1 | - | 4 - 5 | 401 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |