Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006503.1 | NP_001006503.1 | 7 - 8 | 601 | 61 | |
XM_015288626.2 | XP_015144112.1 | 5 - 6 | 778 | 61 | ||
XM_015288627.2 | XP_015144113.1 | 6 - 7 | 780 | 61 | ||
XM_025151603.1 | XP_025007371.1 | 7 - 8 | 614 | 61 | ||
GenBank mRNA | AJ720183.1 | - | 7 - 8 | 601 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |