Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007844.1 | NP_001007845.1 | 8 - 9 | 930 | 103 | |
XM_015295140.2 | XP_015150626.1 | 9 - 10 | 3120 | 103 | ||
XM_015295142.2 | XP_015150628.1 | 7 - 8 | 2871 | 103 | ||
GenBank mRNA | AJ719918.1 | - | 8 - 9 | 930 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |