Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008680.1 | NP_001008680.1 | 1 - 2 | 276 | 88 | |
GenBank mRNA | AJ721100.1 | - | 1 - 2 | 276 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |