Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204135.2 | NP_989466.1 | 4 - 5 | 322 | 101 | |
XM_015289365.2 | XP_015144851.1 | 4 - 5 | 332 | 101 | ||
GenBank mRNA | AF461015.1 | - | 4 - 5 | 330 | 101 | |
AJ719470.1 | - | 4 - 5 | 322 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |