Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204726.1 | NP_990057.1 | 5 - 6 | 363 | 120 | |
XM_015276125.2 | XP_015131611.1 | 5 - 6 | 4762 | 120 | ||
GenBank mRNA | AF082329.1 | - | 5 - 6 | 363 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |