Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144848.2 | NP_001138320.1 | 1 | 1356 | 71 | |
XM_015293823.2 | XP_015149309.2 | 2 | 2619 | 71 | ||
GenBank mRNA | FJ544313.1 | - | 1 | 1351 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |