Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031477.1 | NP_001026648.1 | 2 | 2753 | 94 | |
GenBank mRNA | AJ851534.1 | - | 2 | 2753 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |